Methyl 5-bromo-2-furoate, 98%, Thermo Scientific Chemicals
Methyl 5-bromo-2-furoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 5-bromo-2-furoate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 355750010
Price (USD)/ Each
46.60
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.60
Add to cart
Methyl 5-bromo-2-furoate, 98%, Thermo Scientific Chemicals
Catalog number355750010
Price (USD)/ Each
46.60
-
Add to cart
Chemical Identifiers
IUPAC Namemethyl 5-bromofuran-2-carboxylate
Molecular FormulaC6H5BrO3
InChI KeyFBPIDMAELBIRLE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Br)O1
Molecular Weight (g/mol)205.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Melting point63°C to 68°C
Appearance (Form)Crystalline powder
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only