1x1 image pixel for data collection
Pierce™ 2-Mercaptoethanol
Pierce™ 2-Mercaptoethanol
Thermo Scientific™

Pierce™ 2-Mercaptoethanol

Thermo Scientific Pierce 2-Mercaptoethanol (2-ME), also called beta-mercaptoethanol (b-ME), is a mild reducing agent for cleaving protein disulfide bonds.Features ofRead more
Promo ImagePromotion
Promotion:RELAX25
Reset, recharge, and fuel your next breakthrough
Until September 26, save up to $650 and get a science-inspired break boxLearn More
Have Questions?
Catalog NumberQuantity
35602BIDPromo Image10 x 1 mL
Catalog number 35602BID
Price (USD)
83.65
Online Exclusive
93.25
Save 9.60 (10%)
Each
Promo ImagePromotion available
Add to cart
Quantity:
10 x 1 mL
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
93.25
Save 9.60 (10%)
Each
Add to cart
Thermo Scientific Pierce 2-Mercaptoethanol (2-ME), also called beta-mercaptoethanol (b-ME), is a mild reducing agent for cleaving protein disulfide bonds.

Features of 2-mercaptoethanol:

• Also known as β-mercaptoethanol, beta-mercaptoethanol, bME, b-ME
• Mild but effective reducing agent, often included in enzyme solutions to protect against catalytic site inactivation due to cysteine sulfhydryl oxidation/disulfide formation; added at final concentrations of 5 and 20 mM, with or without EDTA, as an additional protectant

Related Products
Pierce™ TCEP-HCl
Pierce™ DTT (Dithiothreitol)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CAS60-24-2
Concentration14.3 M
FormLiquid
Product LinePierce™
Quantity10 x 1 mL
Product TypeMercaptoethanol
Unit SizeEach
Contents & Storage
Store between 2 to 25°C

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link