Thermo Scientific Chemicals

Phorbol 12-myristate 13-acetate, 97%, Thermo Scientific Chemicals

Catalog number: 356150050
5 mg, Glass bottle, Each
Thermo Scientific Chemicals

Phorbol 12-myristate 13-acetate, 97%, Thermo Scientific Chemicals

Catalog number: 356150050
5 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 356150050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16561-29-8
IUPAC Name
(1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Formula
C36H56O8
InChI Key
PHEDXBVPIONUQT-RGYGYFBISA-N
SMILES
CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
HPLC
>=96.0 %
Appearance (Color)
Colorless to white to light yellow
Appearance (Form)
Film or solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text