1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, 95%, Thermo Scientific Chemicals
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 356190010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS141556-45-8
IUPAC Name1,3-bis(2,4,6-trimethylphenyl)-3H-1λ⁵-imidazol-1-ylium chloride
Molecular FormulaC21H25ClN2
InChI KeyOTOSIXGMLYKKOW-UHFFFAOYSA-M
SMILES[Cl-].CC1=CC(C)=C(N2C=C[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=94 %
Appearance (Color)Off-white to beige
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only