n-Nonanoic acid, 97%, Thermo Scientific Chemicals
n-Nonanoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Nonanoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 356240010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-05-0
IUPAC Namenonanoic acid
Molecular FormulaC9H18O2
InChI KeyFBUKVWPVBMHYJY-UHFFFAOYSA-N
SMILESCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4300 to 1.4340 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only