Thermo Scientific Chemicals

(R)-(-)-2-Methylpiperazine, 98%, ACROS Organics™

Catalog number: 356270050
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-2-Methylpiperazine, 98%, ACROS Organics™

Catalog number: 356270050
5 g, Each
Quantity
Catalog number: 356270050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75336-86-6
IUPAC Name
(2R)-2-methylpiperazine
Molecular Formula
C5H12N2
InChI Key
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
SMILES
C[C@@H]1CNCCN1
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
GC
>=94 %
Melting point
86°C to 91°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text