3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 356480050
Price (USD)/ Each
87.90
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
87.90
Add to cart
3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog number356480050
Price (USD)/ Each
87.90
-
Add to cart
Chemical Identifiers
CAS2888-06-4
IUPAC Name3-chlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4Cl2O2S
InChI KeyOINWZUJVEXUHCC-UHFFFAOYSA-N
SMILESClC1=CC=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5670 to 1.5690 (20°C, 589 nm)
Titration RCOCl>=97.5 %
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only