1-Hexadecyl-3-methylimidazolium chloride monohydrate, 98%, Thermo Scientific Chemicals
1-Hexadecyl-3-methylimidazolium chloride monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Hexadecyl-3-methylimidazolium chloride monohydrate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 356650100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS404001-62-3
IUPAC Name1-hexadecyl-3-methyl-1H-imidazol-3-ium hydrate chloride
Molecular FormulaC20H41ClN2O
InChI KeyYCRGEJFZTMXLMF-UHFFFAOYSA-M
SMILESO.[Cl-].CCCCCCCCCCCCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder and/or chunks
Appearance (Color)Light yellow
Water3 to 5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only