1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 356760050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-methyl-1-octyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC12H23F6N2P
InChI KeyGRCIJNHHTXBJAK-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
Molecular Weight (g/mol)340.29
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)>=98.0 %
Refractive index1.4230 to 1.4250 (20°C, 589 nm)
Water=<1.0 % (K.F.)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only