Thermo Scientific Chemicals

1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Catalog number: 356760250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Catalog number: 356760250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 356760250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
304680-36-2
IUPAC Name
3-methyl-1-octyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular Formula
C12H23F6N2P
InChI Key
GRCIJNHHTXBJAK-UHFFFAOYSA-N
SMILES
F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
NMR (1H-NMR)
>=98.0 %
Refractive index
1.4230 to 1.4250 (20°C, 589 nm)
Water
=<1.0 % (K.F.)
Appearance (Color)
Clear light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text