(R)-2-(Aminomethyl)-1-ethylpyrrolidine, 97%, Thermo Scientific Chemicals
(R)-2-(Aminomethyl)-1-ethylpyrrolidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-2-(Aminomethyl)-1-ethylpyrrolidine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 357000050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-(1-ethylpyrrolidin-2-yl)methanamine
Molecular FormulaC7H16N2
InChI KeyUNRBEYYLYRXYCG-UHFFFAOYNA-N
SMILESCCN1CCCC1CN
Molecular Weight (g/mol)128.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4655 to 1.4685 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only