Thermo Scientific Chemicals

Silica gel, for drying purposes, non-toxic grade, 3-6 mm, Thermo Scientific Chemicals

Catalog number: 357400250
25 kg, Fiber Drum, Each
Thermo Scientific Chemicals

Silica gel, for drying purposes, non-toxic grade, 3-6 mm, Thermo Scientific Chemicals

Catalog number: 357400250
25 kg, Fiber Drum, Each
Quantity
Packaging
Catalog number: 357400250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Form)
Irregular granules
Particle size
3.0 to 6.0 mm
Appearance (Color)
Colorless to white
Residual water
=<2.0 % (140°C, 4 h)
Water adsorption
>=20.0 % (40% rH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent, For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text