Thermo Scientific Chemicals

D-allo-Isoleucine, 97%

Catalog number: 357430010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

D-allo-Isoleucine, 97%

Catalog number: 357430010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 357430010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1509-35-9
IUPAC Name
(2R,3S)-2-amino-3-methylpentanoic acid
Molecular Formula
C6H13NO2
InChI Key
AGPKZVBTJJNPAG-CRCLSJGQSA-N
SMILES
CC[C@H](C)[C@@H](N)C(O)=O
Appearance (Form)
Crystalline powder
Specific optical rotation
-35° to -39° (20°C, 589 nm) (C=5, 1 N HCl)
Infrared spectrum
Conforms
Appearance (Color)
White
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text