Thermo Scientific Chemicals

2,4-Difluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 357490010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Difluorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 357490010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 357490010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13918-92-8
IUPAC Name
2,4-difluorobenzene-1-sulfonyl chloride
Molecular Formula
C6H3ClF2O2S
InChI Key
FJSAJUXIHJIAMD-UHFFFAOYSA-N
SMILES
FC1=CC(F)=C(C=C1)S(Cl)(=O)=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 % (ex Cl)
Refractive index
1.5165 to 1.5185 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text