4-Methylsulphonylbenzyl bromide, Tech., Thermo Scientific Chemicals
4-Methylsulphonylbenzyl bromide, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylsulphonylbenzyl bromide, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number 357580100
Price (USD)/ Each
329.00
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
329.00
Add to cart
4-Methylsulphonylbenzyl bromide, Tech., Thermo Scientific Chemicals
Catalog number357580100
Price (USD)/ Each
329.00
-
Add to cart
Chemical Identifiers
CAS53606-06-7
IUPAC Name1-(chloromethyl)-4-methanesulfonylbenzene
Molecular FormulaC8H9ClO2S
InChI KeyLXPRVXKHIXWBJZ-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=60 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only