Thermo Scientific Chemicals

4-Iodo-3-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: 357800050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Iodo-3-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: 357800050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 357800050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4949-69-3
IUPAC Name
4-iodo-3-methylaniline
Molecular Formula
C7H8IN
InChI Key
UISBOJCPTKUBIC-UHFFFAOYSA-N
SMILES
CC1=CC(N)=CC=C1I
Appearance (Color)
Beige to light brown to dark grey
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
50°C to 55°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text