Thermo Scientific Chemicals

2-Aminoethyl methacrylate hydrochloride, 90%, stabilized, Thermo Scientific Chemicals

Catalog number: 357810050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminoethyl methacrylate hydrochloride, 90%, stabilized, Thermo Scientific Chemicals

Catalog number: 357810050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 357810050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2420-94-2
IUPAC Name
hydrogen 2-aminoethyl 2-methylprop-2-enoate chloride
Molecular Formula
C6H12ClNO2
InChI Key
XSHISXQEKIKSGC-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC(=C)C(=O)OCCN
Appearance (Color)
White to light green to light brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Argentometric
>=88 %
Stabilizer
500 to 1000 ppm phenothiazine

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text