Sodium hydroxide, extra pure, 33wt.% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 33wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, extra pure, 33wt.% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 L
25 L
Packaging:
View all
Plastic Bottle
Catalog number 358625000
Price (USD)/ Each
36.65
Online exclusive
40.90 
Save 4.25 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
40.90 
Save 4.25 (10%)
Add to cart
Sodium hydroxide, extra pure, 33wt.% solution in water, Thermo Scientific Chemicals
Catalog number358625000
Price (USD)/ Each
36.65
Online exclusive
40.90 
Save 4.25 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Density(20°C) 1.355 to 1.365 kg/l
Sodium carbonate (Na2CO3)=<0.1 %
Sodium chloride (NaCl)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only