Thermo Scientific Chemicals

Nickel(II) tetrafluoroborate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 358730250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Nickel(II) tetrafluoroborate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 358730250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 358730250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15684-36-3
IUPAC Name
nickel(2+) bis(tetrafluoroboranuide) hexahydrate
Molecular Formula
B2F8H12NiO6
InChI Key
BFFWGFMAXHCLDV-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F
Appearance (Color)
Blue-green
Appearance (Form)
Powder or crystals
Titration Complexometric
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text