Thermo Scientific Chemicals

4-Aminomethylphenylboronic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: 358842500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

4-Aminomethylphenylboronic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: 358842500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 358842500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75705-21-4
IUPAC Name
[4-(aminomethyl)phenyl]boronic acid hydrochloride
Molecular Formula
C7H11BClNO2
InChI Key
HUZNRXFJHYNUMV-UHFFFAOYSA-N
SMILES
Cl.NCC1=CC=C(C=C1)B(O)O
Titration Argentometric
>=95 %
Appearance (Color)
Light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text