Dibenzothiophene-4-boronic acid, 95%, Thermo Scientific Chemicals
Dibenzothiophene-4-boronic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzothiophene-4-boronic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 359080050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name{8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid
Molecular FormulaC12H9BO2S
InChI KeyGOXNHPQCCUVWRO-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC2=C1SC1=C2C=CC=C1
Molecular Weight (g/mol)228.07
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only