Thermo Scientific Chemicals

3,4-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359090050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359090050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 359090050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151169-75-4
IUPAC Name
(3,4-dichlorophenyl)boronic acid
Molecular Formula
C6H5BCl2O2
InChI Key
JKIGHOARKAIPJI-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(Cl)C(Cl)=C1
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text