Thermo Scientific Chemicals

2-(N,N-Dimethylaminomethyl)phenylboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 359241000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

2-(N,N-Dimethylaminomethyl)phenylboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 359241000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 359241000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
85107-53-5
IUPAC Name
{2-[(dimethylamino)methyl]phenyl}boronic acid
Molecular Formula
C9H14BNO2
InChI Key
VUDCTLOJEPCNRS-UHFFFAOYSA-N
SMILES
CN(C)CC1=CC=CC=C1B(O)O
Appearance (Form)
Powder
Appearance (Color)
Beige
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text