Thermo Scientific Chemicals

3-Formylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Formylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 359370250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87199-16-4
IUPAC Name
(3-formylphenyl)boronic acid
Molecular Formula
C7H7BO3
InChI Key
HJBGZJMKTOMQRR-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC(C=O)=C1
Appearance (Color)
Off-white to beige to light orange
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text