Furan-2-boronic acid, 97%, Thermo Scientific Chemicals
Furan-2-boronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Furan-2-boronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 359390100
Price (USD)/ Each
300.00
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
300.00
Add to cart
Furan-2-boronic acid, 97%, Thermo Scientific Chemicals
Catalog number359390100
Price (USD)/ Each
300.00
-
Add to cart
Chemical Identifiers
CAS13331-23-2
IUPAC Name(furan-2-yl)boronic acid
Molecular FormulaC4H5BO3
InChI KeyPZJSZBJLOWMDRG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only