Thermo Scientific Chemicals

Furan-3-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359400025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Furan-3-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359400025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 359400025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55552-70-0
IUPAC Name
(furan-3-yl)boronic acid
Molecular Formula
C4H5BO3
InChI Key
CYEFKCRAAGLNHW-UHFFFAOYSA-N
SMILES
OB(O)C1=COC=C1
Appearance (Color)
White to yellow to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text