Thermo Scientific Chemicals

3-Hydroxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359410010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Hydroxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359410010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 359410010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87199-18-6
IUPAC Name
(3-hydroxyphenyl)boronic acid
Molecular Formula
C6H7BO3
InChI Key
WFWQWTPAPNEOFE-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC(O)=C1
Appearance (Color)
Pink to grey to tan
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text