Thermo Scientific Chemicals

5-Indolylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359460010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Indolylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 359460010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 359460010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
144104-59-6
IUPAC Name
(1H-indol-5-yl)boronic acid
Molecular Formula
C8H8BNO2
InChI Key
VHADYSUJZAPXOW-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C2NC=CC2=C1
Appearance (Form)
Powder and/or chunks
Titration with NaOH
>=96.0 %
Appearance (Color)
Light yellow to beige
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text