Thermo Scientific Chemicals

Pyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 359650050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Pyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 359650050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 359650050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1692-25-7
IUPAC Name
(pyridin-3-yl)boronic acid
Molecular Formula
C5H6BNO2
InChI Key
ABMYEXAYWZJVOV-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CN=C1
Appearance (Color)
White to yellow to light orange
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text