(2-Methylpropyl)boronic acid, 95+%, Thermo Scientific Chemicals
(2-Methylpropyl)boronic acid, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2-Methylpropyl)boronic acid, 95+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 359950250
Price (USD)/ Each
271.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
271.00
Add to cart
(2-Methylpropyl)boronic acid, 95+%, Thermo Scientific Chemicals
Catalog number359950250
Price (USD)/ Each
271.00
-
Add to cart
Chemical Identifiers
CAS84110-40-7
IUPAC Name(2-methylpropyl)boronic acid
Molecular FormulaC4H11BO2
InChI KeyZAZPDOYUCVFPOI-UHFFFAOYSA-N
SMILESCC(C)CB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH95.0 to 105.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only