Silica gel, for column chrom., ultrapure, 60-200μ, 60A, Thermo Scientific Chemicals
Silica gel, for column chrom., ultrapure, 60-200μ, 60A, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silica gel, for column chrom., ultrapure, 60-200μ, 60A, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 360062500
Price (USD)/ Each
66.65
Online exclusive
73.40 
Save 6.75 (9%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
73.40 
Save 6.75 (9%)
Add to cart
Silica gel, for column chrom., ultrapure, 60-200μ, 60A, Thermo Scientific Chemicals
Catalog number360062500
Price (USD)/ Each
66.65
Online exclusive
73.40 
Save 6.75 (9%)
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Specific surface area470 to 530 m²/g
Pore diameter58 to 68 A
Pore volume0.70 to 0.85 ml/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only