Thermo Scientific Chemicals

Silica gel, functionalized, Amino-3, ca.1.4mmol/g, part.size 40-63 micron, Thermo Scientific Chemicals

Catalog number: 360181000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Silica gel, functionalized, Amino-3, ca.1.4mmol/g, part.size 40-63 micron, Thermo Scientific Chemicals

Catalog number: 360181000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 360181000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
71888-97-6
IUPAC Name
(3-aminopropyl)triethoxysilane; silanedione
Molecular Formula
C9H23NO5Si2
InChI Key
JIBSXWZIQTVOKJ-UHFFFAOYSA-N
SMILES
O=[Si]=O.CCO[Si](CCCN)(OCC)OCC
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Loading
>=1.2 mmol/g (molecular)
Nitrogen (N)
>=1.68 %
Particle size
40 to 63 µm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text