N-α-FMOC-N-Trityl-L-histidine, 98%
N-α-FMOC-N-Trityl-L-histidine, 98%
Thermo Scientific Chemicals

N-α-FMOC-N-Trityl-L-histidine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 360330050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Molecular FormulaC40H33N3O4
InChI KeyXXMYDXUIZKNHDT-UHFFFAOYNA-N
SMILESOC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol)619.72
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Specific optical rotation+80.0° to +89.0° (20°C, 589 nm) (c=1, chloroform) <0.2% D-enantiomer
Appearance of solution(0.5mmol/ml DMF) clear, colourless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only