Thermo Scientific Chemicals

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%, Thermo Scientific Chemicals

Catalog number: 360465000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%, Thermo Scientific Chemicals

Catalog number: 360465000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 360465000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
172843-97-9
IUPAC Name
4-{[(tert-butoxy)carbonyl]amino}oxane-4-carboxylate
Molecular Formula
C11H18NO5
InChI Key
SPPDKPRJPFTBEV-UHFFFAOYSA-M
SMILES
CC(C)(C)OC(=O)NC1(CCOCC1)C([O-])=O
Appearance (Color)
Off-white
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Titration with NaOH
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text