Thermo Scientific Chemicals

2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid, 95%, Thermo Scientific Chemicals

Catalog number: 360485000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid, 95%, Thermo Scientific Chemicals

Catalog number: 360485000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 360485000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
182287-51-0
IUPAC Name
(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(oxan-4-yl)propanoate
Molecular Formula
C13H22NO5
InChI Key
NKYZORHKIYSSEL-SNVBAGLBSA-M
SMILES
CC(C)(C)OC(=O)N[C@H](CC1CCOCC1)C([O-])=O
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
NMR
>=94 %
Appearance (Form)
Chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text