Sodium carbonate, anhydrous, 99.5-100.5% (by anhydrous basis), granular, NF, FCC, Food GMP, J.T.Baker™
Sodium carbonate, anhydrous, 99.5-100.5% (by anhydrous basis), granular, NF, FCC, Food GMP, J.T.Baker™
Avantor™

Sodium carbonate, anhydrous, 99.5-100.5% (by anhydrous basis), granular, NF, FCC, Food GMP, J.T.Baker™

Food GMP Manufactured, meets NF and FCC specificationsRead more
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
3 kg
100 lb.
200 lb.
Catalog number 3605-01
Price (USD)
-
Request A Quote
Quantity:
500 g
Ask our AI about this Product
Food GMP Manufactured, meets NF and FCC specifications
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Melting Point854°C
ColorWhite
pH11.4 to 11.7 (25°C)
Boiling Point1600°C (1013 hPa)
PackagingAmber glass bottle, wide mouth, tamper-evident cap
Quantity500 g
Molecular Weight (g/mol)105.99
Percent Purity99.5 to 100.5%
GradeNF, FCC
Physical FormSolid
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
Certificate of AnalysisFeb 06, 20243605-01
1 result displayed, search above for a specific certificate

Safety Data Sheets

Product Information

Share catalog number, name or link

1x1 image pixel for data collection