4-Bromomethylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
4-Bromomethylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromomethylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 360540050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-(bromomethyl)benzene-1-sulfonyl chloride
Molecular FormulaC7H6BrClO2S
InChI KeyQXTQWYZHHMQSQH-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(CBr)C=C1
Molecular Weight (g/mol)269.54
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point72°C to 77°C
Titration Argentometric>=94 %
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only