3-Bromo-2-chloropyridine, 98%, Thermo Scientific Chemicals
3-Bromo-2-chloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-2-chloropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 360650250
Price (USD)/ Each
134.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
134.00
Add to cart
3-Bromo-2-chloropyridine, 98%, Thermo Scientific Chemicals
Catalog number360650250
Price (USD)/ Each
134.00
-
Add to cart
Chemical Identifiers
IUPAC Name3-bromo-2-chloropyridine
Molecular FormulaC5H3BrClN
InChI KeyHDYNIWBNWMFBDO-UHFFFAOYSA-N
SMILESClC1=NC=CC=C1Br
Molecular Weight (g/mol)192.44
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point53°C to 59°C
Appearance (Form)Crystalline powder or crystals or flakes or solid
GC>=97.5 %
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only