Thermo Scientific Chemicals

1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil, Thermo Scientific Chemicals

Catalog number: 361312500
250 mL, Plastic bottle, Each
Thermo Scientific Chemicals

1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil, Thermo Scientific Chemicals

Catalog number: 361312500
250 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 361312500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3006-86-8, 3006-86-8
Appearance (Form)
Clear liquid
Color scale
=<20 APHA
Titration Iodometric
49 to 51 %
Impurity
=<0.2 % Hydroperoxides (as TBHP)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text