Thermo Scientific Chemicals

(S)-N-BOC-Propargylglycine, 95%, Thermo Scientific Chemicals

Catalog number: 361805000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

(S)-N-BOC-Propargylglycine, 95%, Thermo Scientific Chemicals

Catalog number: 361805000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 361805000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63039-48-5
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}pent-4-ynoate
Molecular Formula
C10H14NO4
InChI Key
AMKHAJIFPHJYMH-ZETCQYMHSA-M
SMILES
CC(C)(C)OC(=O)N[C@@H](CC#C)C([O-])=O
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Specific optical rotation
+24° to +28° (20°C, 589 nm) (C=1, Methanol)
Appearance (Color)
White to light yellow
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text