(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals
(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass Bottle
Catalog number 361830010
Price (USD)/ Each
360.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
360.00
Add to cart
(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals
Catalog number361830010
Price (USD)/ Each
360.00
-
Add to cart
Chemical Identifiers
IUPAC Name(1S,3R)-3-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Molecular FormulaC11H18NO4
InChI KeyRNJQBGXOSAQQDG-JGVFFNPUSA-M
SMILESCC(C)(C)OC(=O)N[C@@H]1CC[C@@H](C1)C([O-])=O
Molecular Weight (g/mol)228.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Appearance (Color)White
Infrared spectrumConforms
Enantiomeric excess>=97.5 %
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only