(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, 98% ee, Thermo Scientific Chemicals
(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, 98% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one, 95%, 98% ee, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 362200010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(1S,2R,6S,7R)-4,4-dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0²,⁶]decan-9-one
Molecular FormulaC9H13NO3
InChI KeyCSGIROCFQXHTKS-WNJXEPBRSA-N
SMILESCC1(C)O[C@H]2[C@H]3C[C@@H]([C@H]2O1)C(=O)N3
Molecular Weight (g/mol)183.21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals or needles
Enantiomeric excess>=97.5 % (GC)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only