Thermo Scientific Chemicals

(1R,3S)-3-Aminocyclopentanecarboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals

Catalog number: 362270010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

(1R,3S)-3-Aminocyclopentanecarboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals

Catalog number: 362270010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 362270010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
71830-08-5
IUPAC Name
(1R,3S)-3-azaniumylcyclopentane-1-carboxylate
Molecular Formula
C6H11NO2
InChI Key
MLLSSTJTARJLHK-UHNVWZDZSA-N
SMILES
[NH3+][C@H]1CC[C@H](C1)C([O-])=O
Appearance (Form)
Crystalline powder
HPLC
>=94 %
Appearance (Color)
Off-white to beige
Enantiomeric excess
>=97.5 % (HPLC)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text