(1S,3R)-3-Aminocyclopentanecarboxylic acid, 95%, >90% ee, Thermo Scientific Chemicals
(1S,3R)-3-Aminocyclopentanecarboxylic acid, 95%, >90% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1S,3R)-3-Aminocyclopentanecarboxylic acid, 95%, >90% ee, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 362280010
Price (USD)/ Each
450.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
450.00
Add to cart
(1S,3R)-3-Aminocyclopentanecarboxylic acid, 95%, >90% ee, Thermo Scientific Chemicals
Catalog number362280010
Price (USD)/ Each
450.00
-
Add to cart
Chemical Identifiers
CAS71830-07-4
IUPAC Name(1S,3R)-3-azaniumylcyclopentane-1-carboxylate
Molecular FormulaC6H11NO2
InChI KeyMLLSSTJTARJLHK-CRCLSJGQSA-N
SMILES[NH3+][C@@H]1CC[C@@H](C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration with HClO4>=94 %
Infrared spectrumConforms
Enantiomeric excess>=90 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only