(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals
(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 362530010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS220497-88-1
IUPAC Name(1S,2R,3S,4S)-2,3-dihydroxy-4-(hydroxymethyl)cyclopentan-1-aminium
Molecular FormulaC6H14NO3
InChI KeyFHNKBDPGQXLKRW-OMMKOOBNSA-O
SMILES[NH3+][C@H]1C[C@@H](CO)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
Appearance (Color)White to off-white
Infrared spectrumConforms
Enantiomeric excess>=97.5 %
HPLC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only