(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 362990010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS340188-50-3
IUPAC Name(1R)-3-ethoxy-3-oxo-1-phenylpropan-1-aminium
Molecular FormulaC11H16NO2
InChI KeyNUWRDXMXYDWUAN-SNVBAGLBSA-O
SMILESCCOC(=O)C[C@@H]([NH3+])C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Enantiomeric excess>=97.5 % (GC)
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=94 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only