Thermo Scientific Chemicals

1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 363230250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 363230250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 363230250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14647-23-5
IUPAC Name
[2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel
Molecular Formula
C26H24Cl2NiP2
InChI Key
XXECWTBMGGXMKP-UHFFFAOYSA-L
SMILES
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Orange to red
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Elemental Analysis
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text