Acetylacetonatobis(ethylene)rhodium(I), 99%, Thermo Scientific Chemicals
Acetylacetonatobis(ethylene)rhodium(I), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetylacetonatobis(ethylene)rhodium(I), 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number 363381000
Price (USD)/ Each
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12082-47-2
IUPAC Nameλ¹-rhodium(1+) bis(ethene) (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC9H15O2Rh
InChI KeyFLRBEQQDEGBCJS-FGSKAQBVSA-M
SMILES[Rh+].C=C.C=C.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Elemental Analysis>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only