Thermo Scientific Chemicals

(Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate, Thermo Scientific Chemicals

Catalog number: 363581000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

(Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate, Thermo Scientific Chemicals

Catalog number: 363581000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 363581000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64536-78-3
IUPAC Name
iridium(1+) (1Z,5Z)-cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide pyridine tricyclohexylphosphane
Molecular Formula
C31H50F6IrNP2
InChI Key
UJXHUUQZACSUOG-KJWGIZLLSA-N
SMILES
[Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Appearance (Color)
Orange
Infrared spectrum
Conforms
Carbon (C)
45.6 to 47.0 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text