Thermo Scientific Chemicals

Dichloro(1,5-cyclooctadiene)platinum(II), 98%, Thermo Scientific Chemicals

Catalog number: 363612500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

Dichloro(1,5-cyclooctadiene)platinum(II), 98%, Thermo Scientific Chemicals

Catalog number: 363612500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 363612500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12080-32-9
IUPAC Name
(1Z,5Z)-cycloocta-1,5-diene; dichloroplatinum
Molecular Formula
C8H12Cl2Pt
InChI Key
VVAOPCKKNIUEEU-PHFPKPIQSA-L
SMILES
Cl[Pt]Cl.C1C\C=C/CC\C=C/1
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Assay
>=97.5 % (ex Pt)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text